Product Details

SNP ID
rs3829815
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:7122913 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGAGCATGTGGTAGGATCAGTATC[A/G]CAAGCCCCTAGTTCTTGGCCAGAGC
Phenotype
MIM: 611324 MIM: 611866
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
C1RL-AS1 PubMed Links

Gene Details

Gene
C1RL-AS1
Gene Name
C1RL antisense RNA 1
There are no transcripts associated with this gene.

Gene
CLSTN3
Gene Name
calsyntenin 3
There are no transcripts associated with this gene.

Gene
RBP5
Gene Name
retinol binding protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017020002.1 Intron XP_016875491.1

View Full Product Details