Product Details

SNP ID
rs1167828
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:75534076 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AAATGGAGCCAGGAGACCCCTCTAG[A/G]CTAAGGAGCTAAGAGGGAACTTCCA
Phenotype
MIM: 601767
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HIP1 PubMed Links

Gene Details

Gene
HIP1
Gene Name
huntingtin interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243198.2 7268 UTR 3 NP_001230127.1
NM_005338.6 7268 UTR 3 NP_005329.3
XM_005250304.2 7268 UTR 3 XP_005250361.1
XM_005250305.2 7268 UTR 3 XP_005250362.1
XM_011516116.2 7268 UTR 3 XP_011514418.1
XM_017012099.1 7268 UTR 3 XP_016867588.1
Gene
PMS2P3
Gene Name
PMS1 homolog 2, mismatch repair system component pseudogene 3
There are no transcripts associated with this gene.

View Full Product Details