Product Details

SNP ID
rs61902821
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:58190488 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCGTTGCAGGTGAACCACTGGA[C/T]GGATGGCTGAAAGGAATTACACCGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR9Q1 PubMed Links

Gene Details

Gene
OR9Q1
Gene Name
olfactory receptor family 9 subfamily Q member 1
There are no transcripts associated with this gene.

Gene
OR9Q2
Gene Name
olfactory receptor family 9 subfamily Q member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005283.2 55 UTR 5 NP_001005283.1

View Full Product Details