Product Details

SNP ID
rs12421556
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:58041164 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
TGTGGGCCAGGGTTCTGTAGAGTTT[C/T]ACAGGCTGAGGGGGCCATCTGGTCA
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
OR6Q1 PubMed Links
Additional Information
For this assay, SNP(s) [rs77165873] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
OR6Q1
Gene Name
olfactory receptor family 6 subfamily Q member 1 (gene/pseudogene)
There are no transcripts associated with this gene.

Gene
OR9Q1
Gene Name
olfactory receptor family 9 subfamily Q member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005212.3 Intron NP_001005212.1
Gene
VN2R9P
Gene Name
vomeronasal 2 receptor 9 pseudogene
There are no transcripts associated with this gene.

View Full Product Details