Product Details
- SNP ID
-
rs78910164
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:77703286 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATGAATGAGATATTCATCTTCCAC[A/G]TGTAATACCAAGACCCCAGACAGCT
- Phenotype
-
MIM: 610491
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MIR3665
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77310582] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MIR3665
- Gene Name
- microRNA 3665
There are no transcripts associated with this gene.
- Gene
- SLAIN1
- Gene Name
- SLAIN motif family member 1
View Full Product Details