Product Details

SNP ID
rs7109706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:31439973 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCTCAGTCTCTTTAAGTTTCACC[A/G]TAAGCAGGCACAATTTAGTATTGAG
Phenotype
MIM: 611072 MIM: 612323
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC24 PubMed Links

Gene Details

Gene
DNAJC24
Gene Name
DnaJ heat shock protein family (Hsp40) member C24
There are no transcripts associated with this gene.

Gene
IMMP1L
Gene Name
inner mitochondrial membrane peptidase subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304274.1 Intron NP_001291203.1
NM_144981.2 Intron NP_659418.1
XM_005252812.3 Intron XP_005252869.1
XM_011519942.1 Intron XP_011518244.1
XM_011519943.2 Intron XP_011518245.1
XM_011519944.2 Intron XP_011518246.1
XM_011519945.2 Intron XP_011518247.1
XM_011519946.2 Intron XP_011518248.1
XM_011519947.2 Intron XP_011518249.1
XM_017017305.1 Intron XP_016872794.1
XM_017017306.1 Intron XP_016872795.1
XM_017017307.1 Intron XP_016872796.1

View Full Product Details