Product Details
- SNP ID
-
rs3780178
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:75093236 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CCTTTTTCTGAATGAGTACAACCTC[C/T]TAGTTTTACACATATAGCATTTATG
- Phenotype
-
MIM: 608704
MIM: 610180
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NMRK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs115434887] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NMRK1
- Gene Name
- nicotinamide riboside kinase 1
There are no transcripts associated with this gene.
- Gene
- OSTF1
- Gene Name
- osteoclast stimulating factor 1
View Full Product Details