Product Details

SNP ID
rs4644918
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:80035232 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTAGCGTCGGACTTTAGCTTCCA[A/T]CGTGTTCCAGTTAATGAGCCGATAT
Phenotype
MIM: 611595
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RBFA PubMed Links

Gene Details

Gene
RBFA
Gene Name
ribosome binding factor A (putative)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171967.1 Intron NP_001165438.1
NM_024805.2 Intron NP_079081.2
Gene
TXNL4A
Gene Name
thioredoxin like 4A
There are no transcripts associated with this gene.

View Full Product Details