Product Details

SNP ID
rs11873739
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:10461221 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGCCCTCCAGAGCTCCAGGATCT[C/T]ATTTAGTGCTCTGGATATGGTTGTC
Phenotype
MIM: 607479
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APCDD1 PubMed Links

Gene Details

Gene
APCDD1
Gene Name
APC down-regulated 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153000.4 Intron NP_694545.1
XM_011525603.1 Intron XP_011523905.1

View Full Product Details