Product Details

SNP ID
rs11212484
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:108013664 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGAAGGCAGTAGTATATAACCGAAC[A/C]ACAAGATAAATCAAACTTTAAACTG
Phenotype
MIM: 601741
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CUL5 PubMed Links

Gene Details

Gene
CUL5
Gene Name
cullin 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003478.3 Intron NP_003469.2
XM_005271682.2 Intron XP_005271739.1
XM_011543013.1 Intron XP_011541315.1
XM_017018363.1 Intron XP_016873852.1
XM_017018364.1 Intron XP_016873853.1
XM_017018365.1 Intron XP_016873854.1

View Full Product Details