Product Details
- SNP ID
-
rs4753827
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:108016603 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AGTGTCCTGGTGAACCCAGTTGTCA[G/T]TAAGAGCCATTTGCCTCCTGCCTTG
- Phenotype
-
MIM: 601741
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CUL5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74735899] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CUL5
- Gene Name
- cullin 5
View Full Product Details