Product Details

SNP ID
rs3793137
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:39904728 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGACTTCTACCAGGGATGCCTTCA[A/C]GCCAAGGCTGTTCTCACCAGCTGCC
Phenotype
MIM: 606627 MIM: 603707
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DAAM2 PubMed Links

Gene Details

Gene
DAAM2
Gene Name
dishevelled associated activator of morphogenesis 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001201427.1 3537 UTR 3 NP_001188356.1
NM_015345.3 3537 UTR 3 NP_056160.2
XM_006715039.3 3537 UTR 3 XP_006715102.3
XM_006715040.3 3537 Intron XP_006715103.3
XM_006715042.2 3537 UTR 3 XP_006715105.1
XM_006715043.2 3537 UTR 3 XP_006715106.1
XM_006715045.3 3537 UTR 3 XP_006715108.1
XM_006715046.3 3537 UTR 3 XP_006715109.1
XM_017010630.1 3537 UTR 3 XP_016866119.1
Gene
LOC100505635
Gene Name
uncharacterized LOC100505635
There are no transcripts associated with this gene.

Gene
MOCS1
Gene Name
molybdenum cofactor synthesis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001075098.3 3537 UTR 3 NP_001068566.1
NM_005943.5 3537 UTR 3 NP_005934.2
XM_011514632.2 3537 Intron XP_011512934.1
XM_011514633.2 3537 UTR 3 XP_011512935.1
XM_011514634.2 3537 Intron XP_011512936.1
XM_017010888.1 3537 UTR 3 XP_016866377.1

View Full Product Details