Product Details
- SNP ID
-
rs13279074
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:64581107 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTGGGTGTCCCCGGGCTGCTAGTAG[G/T]TTCAGCCGGCGTTGGGGGCGACCCT
- Phenotype
-
MIM: 613483
MIM: 603711
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BHLHE22
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs544639534] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BHLHE22
- Gene Name
- basic helix-loop-helix family member e22
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152414.4 |
870 |
Missense Mutation |
GGT,GTT |
G106V |
NP_689627.1 |
- Gene
- CYP7B1
- Gene Name
- cytochrome P450 family 7 subfamily B member 1
There are no transcripts associated with this gene.
- Gene
- LOC401463
- Gene Name
- uncharacterized LOC401463
There are no transcripts associated with this gene.
View Full Product Details