Product Details

SNP ID
rs12674515
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:38280012 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGATTAGAGATCTCAAAGTTCTG[A/G]TAACTGCTCTGTTGTTTTGCTGTCA
Phenotype
MIM: 615003 MIM: 607083
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDHD2 PubMed Links

Gene Details

Gene
DDHD2
Gene Name
DDHD domain containing 2
There are no transcripts associated with this gene.

Gene
WHSC1L1
Gene Name
Wolf-Hirschhorn syndrome candidate 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017778.2 Intron NP_060248.2
NM_023034.1 Intron NP_075447.1

View Full Product Details