Product Details

SNP ID
rs7176482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:52311166 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGACGCTTCAGTACATGAGCTGTCC[A/G]GAGTAAAAAAAAGGGCTGCATGGCA
Phenotype
MIM: 160777
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MYO5A PubMed Links

Gene Details

Gene
MYO5A
Gene Name
myosin VA
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000259.3 8144 UTR 3 NP_000250.3
NM_001142495.1 8144 UTR 3 NP_001135967.1
XM_005254397.3 8144 UTR 3 XP_005254454.1
XM_011521606.2 8144 UTR 3 XP_011519908.2
XM_011521607.2 8144 UTR 3 XP_011519909.2
XM_011521608.2 8144 UTR 3 XP_011519910.2
XM_011521609.2 8144 UTR 3 XP_011519911.2
XM_011521610.2 8144 UTR 3 XP_011519912.2
XM_011521611.2 8144 UTR 3 XP_011519913.2
XM_011521612.2 8144 UTR 3 XP_011519914.2
XM_017022227.1 8144 UTR 3 XP_016877716.1

View Full Product Details