Product Details

SNP ID
rs7937488
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:86382359 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAATGAATGAGGGGCAACTTGGAG[A/G]TAAAATCTACAGATTATGGTGATGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC81 PubMed Links

Gene Details

Gene
CCDC81
Gene Name
coiled-coil domain containing 81
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001156474.1 Intron NP_001149946.1
NM_021827.4 Intron NP_068599.3
XM_005274161.1 Intron XP_005274218.1
XM_005274162.3 Intron XP_005274219.1
XM_017018105.1 Intron XP_016873594.1

View Full Product Details