Product Details

SNP ID
rs6014978
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:57332336 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAGCAAACCACTTGCTCAGGGGAA[A/G]TATAACCAATCCTGATACCAGACCA
Phenotype
MIM: 605114
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPO11 PubMed Links

Gene Details

Gene
SPO11
Gene Name
SPO11, initiator of meiotic double stranded breaks
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012444.2 Intron NP_036576.1
NM_198265.1 Intron NP_937998.1
XM_005260379.3 Intron XP_005260436.1
XM_005260380.3 Intron XP_005260437.1
XM_005260381.3 Intron XP_005260438.1
XM_005260382.4 Intron XP_005260439.1
XM_011528756.2 Intron XP_011527058.1
XM_011528757.2 Intron XP_011527059.1

View Full Product Details