Product Details
- SNP ID
-
rs35389724
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:128901376 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACGTGTAAGGCACCCAGTCCAGAG[C/T]AGTGCACGCAAACACACACACACAC
- Phenotype
-
MIM: 600734
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
C11orf45
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs144517706] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- C11orf45
- Gene Name
- chromosome 11 open reading frame 45
- Gene
- KCNJ5
- Gene Name
- potassium voltage-gated channel subfamily J member 5
View Full Product Details