Product Details

SNP ID
rs8063688
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:80603118 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAGATGAGTCATTTCAGCTTCTGG[A/G]GGGAAAAGATTCAGACCTCCAGACA
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
CDYL2 PubMed Links
Additional Information
For this assay, SNP(s) [rs193109758] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
CDYL2
Gene Name
chromodomain protein, Y-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152342.3 3451 UTR 3 NP_689555.2
XM_011522866.1 3451 UTR 3 XP_011521168.1
XM_011522867.2 3451 UTR 3 XP_011521169.1

View Full Product Details