Product Details

SNP ID
rs10891301
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:111938765 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGGCCCCCGTGCCCTAGCGTTGTTC[C/T]TAACACAGAGGAAAACTGCAGAAAT
Phenotype
MIM: 610493
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DIXDC1 PubMed Links

Gene Details

Gene
DIXDC1
Gene Name
DIX domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037954.3 Intron NP_001033043.1
NM_001278542.1 Intron NP_001265471.1
NM_033425.4 Intron NP_219493.1
XM_017018466.1 Intron XP_016873955.1
XM_017018467.1 Intron XP_016873956.1
XM_017018468.1 Intron XP_016873957.1
XM_017018469.1 Intron XP_016873958.1

View Full Product Details