Product Details
- SNP ID
-
rs11636944
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:99107400 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- CATGGAGATTTGACCAGTGGTTATT[G/T]TGACTCTCCAAAAAGGAGTATCTGT
- Phenotype
-
MIM: 606087
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SYNM
PubMed Links
Gene Details
- Gene
- SYNM
- Gene Name
- synemin
View Full Product Details