Product Details

SNP ID
rs260089
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.15:99137940 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GACTGTTGAATTCCATTTTCTAGGC[A/G]GAGGTGATTTGTACAGCACCCTAAA
Phenotype
MIM: 606087
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SYNM PubMed Links

Gene Details

Gene
SYNM
Gene Name
synemin
There are no transcripts associated with this gene.

Gene
TTC23
Gene Name
tetratricopeptide repeat domain 23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288615.1 1806 UTR 3 NP_001275544.1
NM_001288616.1 1806 UTR 3 NP_001275545.1
XM_011521927.2 1806 UTR 3 XP_011520229.1
XM_011521929.1 1806 UTR 3 XP_011520231.1
XM_011521930.1 1806 UTR 3 XP_011520232.1
XM_011521931.1 1806 UTR 3 XP_011520233.1
XM_011521932.1 1806 UTR 3 XP_011520234.1
XM_011521933.1 1806 UTR 3 XP_011520235.1
XM_011521934.1 1806 UTR 3 XP_011520236.1
XM_011521935.1 1806 UTR 3 XP_011520237.1
XM_011521936.1 1806 UTR 3 XP_011520238.1
XM_011521937.1 1806 UTR 3 XP_011520239.1
XM_011521939.1 1806 UTR 3 XP_011520241.1
XM_011521940.2 1806 UTR 3 XP_011520242.1
XM_011521941.2 1806 UTR 3 XP_011520243.1
XM_011521943.2 1806 Intron XP_011520245.1
XM_011521944.2 1806 Intron XP_011520246.1
XM_011521945.2 1806 Intron XP_011520247.1
XM_017022509.1 1806 UTR 3 XP_016877998.1
XM_017022510.1 1806 UTR 3 XP_016877999.1
XM_017022511.1 1806 UTR 3 XP_016878000.1
XM_017022512.1 1806 UTR 3 XP_016878001.1
XM_017022513.1 1806 UTR 3 XP_016878002.1
XM_017022514.1 1806 UTR 3 XP_016878003.1
XM_017022515.1 1806 UTR 3 XP_016878004.1
XM_017022516.1 1806 UTR 3 XP_016878005.1
XM_017022517.1 1806 UTR 3 XP_016878006.1
XM_017022518.1 1806 UTR 3 XP_016878007.1

View Full Product Details