Product Details

SNP ID
rs958584
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:29063206 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTGGGTGGCTGGACTTCCCTGCGTT[A/G]CCTGCAGTCCAGGCAGCTCCTAAGT
Phenotype
MIM: 613425
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf71 PubMed Links

Gene Details

Gene
C2orf71
Gene Name
chromosome 2 open reading frame 71
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001029883.2 5530 UTR 3 NP_001025054.1

View Full Product Details