Product Details

SNP ID
rs13281311
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:28023867 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTTTCTGGCATCTCAGGTTTTAA[A/T]CCCATGATCTGTAATCAAACCATGT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NUGGC PubMed Links

Gene Details

Gene
NUGGC
Gene Name
nuclear GTPase, germinal center associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001010906.1 Intron NP_001010906.1
XM_011544523.2 Intron XP_011542825.1
XM_011544524.2 Intron XP_011542826.1
XM_011544525.1 Intron XP_011542827.1
XM_011544526.2 Intron XP_011542828.1
XM_017013403.1 Intron XP_016868892.1

View Full Product Details