Product Details

SNP ID
rs11986256
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:30736352 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGATCTCTTTCACGCTTTTGATCTC[A/G]TTTCAGTAACATATCTTGAAAGATC
Phenotype
MIM: 138300 MIM: 602155
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSR PubMed Links

Gene Details

Gene
GSR
Gene Name
glutathione-disulfide reductase
There are no transcripts associated with this gene.

Gene
LOC105379356
Gene Name
uncharacterized LOC105379356
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017014155.1 Intron XP_016869644.1
XM_017014156.1 Intron XP_016869645.1
Gene
UBXN8
Gene Name
UBX domain protein 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282189.1 Intron NP_001269118.1
NM_001282199.1 Intron NP_001269128.1
NM_005671.3 Intron NP_005662.1
XM_011544655.2 Intron XP_011542957.1
XM_017013862.1 Intron XP_016869351.1

View Full Product Details