Product Details

SNP ID
rs10152118
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:69836657 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTCATCCCTCACCCTTCATGGGA[C/T]GTGTGTCTGAGTGGGGTTGGGGGAC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LINC00593 PubMed Links

Gene Details

Gene
LINC00593
Gene Name
long intergenic non-protein coding RNA 593
There are no transcripts associated with this gene.

View Full Product Details