Product Details

SNP ID
rs989655
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:37252711 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGCACCCAGTTCTCCTCCATGAGG[G/T]TATTCTCTCCATCTCCAGCTTTTCG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NWD2 PubMed Links

Gene Details

Gene
NWD2
Gene Name
NACHT and WD repeat domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144990.1 Intron NP_001138462.1

View Full Product Details