Product Details

SNP ID
rs6879260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:180304014 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAGTGGGGCTGGCGACTGAGTT[C/T]GATCACATGGCCAATGGTTCACTCC
Phenotype
MIM: 603865
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GFPT2 PubMed Links

Gene Details

Gene
GFPT2
Gene Name
glutamine-fructose-6-phosphate transaminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005110.3 Intron NP_005101.1
XM_006714942.2 Intron XP_006715005.1
XM_017010100.1 Intron XP_016865589.1

View Full Product Details