Product Details
- SNP ID
-
rs2047589
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:80320725 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGACTTGACAAAATCAATGAAAT[A/G]TTATCAACAAACCTGCCTGTTAGTT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SPZ1
PubMed Links
Gene Details
- Gene
- SPZ1
- Gene Name
- spermatogenic leucine zipper 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032567.3 |
755 |
Missense Mutation |
ATA,ATG |
I170M |
NP_115956.3 |
View Full Product Details