Product Details

SNP ID
rs6601073
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179750270 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTTGCTTAGACTTCACATGCTTA[C/T]TCTGTGAGCCCTTTCCTGAGCACCT
Phenotype
MIM: 605424
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAML1 PubMed Links

Gene Details

Gene
MAML1
Gene Name
mastermind like transcriptional coactivator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014757.4 Intron NP_055572.1

View Full Product Details