Product Details

SNP ID
rs35289294
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179814526 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAATAAAAGTATACTCAAAGCTGTA[C/T]ACCCTTCAAGGGTCCCCTGGGGTCT
Phenotype
MIM: 604561 MIM: 601530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MGAT4B PubMed Links

Gene Details

Gene
MGAT4B
Gene Name
mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B
There are no transcripts associated with this gene.

Gene
SQSTM1
Gene Name
sequestosome 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142298.1 2743 Intron NP_001135770.1
NM_001142299.1 2743 Intron NP_001135771.1
NM_003900.4 2743 Intron NP_003891.1
XM_011534683.2 2743 UTR 3 XP_011532985.1
XM_017010010.1 2743 Intron XP_016865499.1

View Full Product Details