Product Details

SNP ID
rs35647750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:179817132 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGGGGTCAGCGCTGGCGTCGCC[A/G]CTCGGGTGGGGTGGGCCGGGTTCGG
Phenotype
MIM: 601530
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SQSTM1 PubMed Links

Gene Details

Gene
SQSTM1
Gene Name
sequestosome 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142298.1 Intron NP_001135770.1
NM_001142299.1 Intron NP_001135771.1
NM_003900.4 Intron NP_003891.1
XM_011534683.2 Intron XP_011532985.1
XM_017010010.1 Intron XP_016865499.1

View Full Product Details