Product Details

SNP ID
rs13011076
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:222577581 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGCAGGGTATACTATTACAAATCC[A/G]GACAGGATCCTGTAGGACCTTATGC
Phenotype
MIM: 609690
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FARSB PubMed Links

Gene Details

Gene
FARSB
Gene Name
phenylalanyl-tRNA synthetase beta subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005687.4 Intron NP_005678.3
XM_006712169.2 Intron XP_006712232.1
XM_011510466.2 Intron XP_011508768.1
XM_017003110.1 Intron XP_016858599.1

View Full Product Details