Product Details

SNP ID
rs411245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:154046393 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATCAAATTTAAACTAATCTCCAT[A/C]AATATTTATTGAGCATGTGCTAGTT
Phenotype
MIM: 600491
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM114A2 PubMed Links

Gene Details

Gene
FAM114A2
Gene Name
family with sequence similarity 114 member A2
There are no transcripts associated with this gene.

Gene
MFAP3
Gene Name
microfibrillar associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135037.1 Intron NP_001128509.1
NM_001242336.1 Intron NP_001229265.1
NM_005927.4 Intron NP_005918.1

View Full Product Details