Product Details

SNP ID
rs4690219
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:983482 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCGTTGTGCCCCCACCCAGCAGGA[A/G]GGAGAGCTGCAGAGAGAGGCCTCCA
Phenotype
MIM: 601207 MIM: 252800 MIM: 610130
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DGKQ PubMed Links

Gene Details

Gene
DGKQ
Gene Name
diacylglycerol kinase theta
There are no transcripts associated with this gene.

Gene
IDUA
Gene Name
iduronidase, alpha-L-
There are no transcripts associated with this gene.

Gene
SLC26A1
Gene Name
solute carrier family 26 member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022042.3 Intron NP_071325.2
NM_134425.2 Intron NP_602297.1
NM_213613.3 Intron NP_998778.1

View Full Product Details