Product Details

SNP ID
rs4783949
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56677497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAGCAAGTTTTTCACAGGAAGGCT[C/G]TCACTCCAAAGATCCACCATTGTCT
Phenotype
MIM: 156353 MIM: 156354 MIM: 156355 MIM: 156359
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MT1G PubMed Links

Gene Details

Gene
MT1G
Gene Name
metallothionein 1G
There are no transcripts associated with this gene.

Gene
MT1H
Gene Name
metallothionein 1H
There are no transcripts associated with this gene.

Gene
MT1IP
Gene Name
metallothionein 1I, pseudogene
There are no transcripts associated with this gene.

Gene
MT1X
Gene Name
metallothionein 1X
There are no transcripts associated with this gene.

View Full Product Details