Product Details

SNP ID
rs1854574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34726674 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTAATCTTGTATGCCATCTGCATAC[A/G]TTGACTGGGCAGCTGACTGGGTTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM205A PubMed Links

Gene Details

Gene
FAM205A
Gene Name
family with sequence similarity 205 member A
There are no transcripts associated with this gene.

View Full Product Details