Product Details

SNP ID
rs10981994
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:113887187 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATACCTAGACAGACTGTAGGACTCA[A/G]AACCTGGCATGGGATGGTGATGTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF618 PubMed Links

Gene Details

Gene
ZNF618
Gene Name
zinc finger protein 618
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318040.1 Intron NP_001304969.1
NM_001318041.1 Intron NP_001304970.1
NM_001318042.1 Intron NP_001304971.1
NM_133374.2 Intron NP_588615.2
XM_005251692.2 Intron XP_005251749.1
XM_005251699.2 Intron XP_005251756.1
XM_011518200.1 Intron XP_011516502.1
XM_011518201.1 Intron XP_011516503.1
XM_011518203.2 Intron XP_011516505.1
XM_011518204.2 Intron XP_011516506.1
XM_011518209.2 Intron XP_011516511.1
XM_017014241.1 Intron XP_016869730.1
XM_017014242.1 Intron XP_016869731.1
XM_017014243.1 Intron XP_016869732.1
XM_017014244.1 Intron XP_016869733.1
XM_017014245.1 Intron XP_016869734.1
XM_017014246.1 Intron XP_016869735.1
XM_017014247.1 Intron XP_016869736.1
XM_017014248.1 Intron XP_016869737.1
XM_017014249.1 Intron XP_016869738.1
XM_017014250.1 Intron XP_016869739.1
XM_017014251.1 Intron XP_016869740.1
XM_017014252.1 Intron XP_016869741.1

View Full Product Details