Product Details

SNP ID
rs12685583
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122191146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGGTTGGAAGTGCCTCTGCTC[C/T]CTGCACGTGATGACACACACTGAGA
Phenotype
MIM: 608215
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LHX6 PubMed Links

Gene Details

Gene
LHX6
Gene Name
LIM homeobox 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242333.1 Intron NP_001229262.1
NM_001242334.1 Intron NP_001229263.1
NM_001242335.1 Intron NP_001229264.1
NM_014368.4 Intron NP_055183.2
NM_199160.3 Intron NP_954629.2
XM_005251916.3 Intron XP_005251973.1
XM_011518521.2 Intron XP_011516823.1
XM_011518522.2 Intron XP_011516824.1
Gene
MORN5
Gene Name
MORN repeat containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286828.1 Intron NP_001273757.1
NM_198469.3 Intron NP_940871.2
XM_005251877.4 Intron XP_005251934.1
XM_005251878.3 Intron XP_005251935.1
XM_006717034.3 Intron XP_006717097.1
XM_011518457.2 Intron XP_011516759.1
XM_011518458.1 Intron XP_011516760.1
XM_017014578.1 Intron XP_016870067.1

View Full Product Details