Product Details

SNP ID
rs2183424
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:181036798 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGTATACCTAGTGCCCAACAG[A/G]CACTTAGTAAATGTTTGCTGAGTGA
Phenotype
MIM: 600764
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MR1 PubMed Links

Gene Details

Gene
MR1
Gene Name
major histocompatibility complex, class I-related
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001194999.1 Intron NP_001181928.1
NM_001195000.1 Intron NP_001181929.1
NM_001195035.1 Intron NP_001181964.1
NM_001310213.1 Intron NP_001297142.1
NM_001531.2 Intron NP_001522.1
XM_006711286.3 Intron XP_006711349.1
XM_011509463.2 Intron XP_011507765.1

View Full Product Details