Product Details

SNP ID
rs2868364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46029657 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACGGCTTCGGGCTGACTTGGGTTA[A/G]CTAAACCGAGGAGGCGGTGGGAAGG
Phenotype
MIM: 606726
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC12A5 PubMed Links

Gene Details

Gene
SLC12A5
Gene Name
solute carrier family 12 member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134771.1 Intron NP_001128243.1
NM_020708.4 Intron NP_065759.1
XM_017027981.1 Intron XP_016883470.1

View Full Product Details