Product Details

SNP ID
rs34356435
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.3:15606817 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
GGCCAAGGAGCAAGTCCTTTGCAAT[C/G]CAATTGTAATTACAGCCAGCTTGTC
Phenotype
MIM: 609019 MIM: 604300
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BTD PubMed Links

Gene Details

Gene
BTD
Gene Name
biotinidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001281726.1 Intron NP_001268655.1
XM_006713314.3 Intron XP_006713377.1
XM_011534041.2 Intron XP_011532343.1
XM_017007088.1 Intron XP_016862577.1
Gene
HACL1
Gene Name
2-hydroxyacyl-CoA lyase 1
There are no transcripts associated with this gene.

View Full Product Details