Product Details

SNP ID
rs12592252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:75940264 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACCATAGCGTCAAAAAGAATAAAA[C/T]GCACAGAAATTAGCTTACCCAAGGT
Phenotype
MIM: 609096 MIM: 610894
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FBXO22 PubMed Links

Gene Details

Gene
FBXO22
Gene Name
F-box protein 22
There are no transcripts associated with this gene.

Gene
FBXO22-AS1
Gene Name
FBXO22 antisense RNA 1
There are no transcripts associated with this gene.

Gene
NRG4
Gene Name
neuregulin 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138573.3 6999 Intron NP_612640.1
XM_017021937.1 6999 Intron XP_016877426.1
XM_017021938.1 6999 Intron XP_016877427.1
XM_017021939.1 6999 Intron XP_016877428.1
XM_017021940.1 6999 Intron XP_016877429.1
XM_017021941.1 6999 Intron XP_016877430.1
XM_017021942.1 6999 Intron XP_016877431.1
XM_017021943.1 6999 Intron XP_016877432.1
XM_017021944.1 6999 Intron XP_016877433.1
XM_017021945.1 6999 UTR 3 XP_016877434.1
XM_017021946.1 6999 Intron XP_016877435.1
XM_017021947.1 6999 UTR 3 XP_016877436.1
XM_017021948.1 6999 UTR 3 XP_016877437.1

View Full Product Details