Product Details

SNP ID
rs11820182
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64042843 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCACCAGGCTTGGGCTAGCCAGAG[A/G]GGCTGGGTAGTGCCACACCCCAGTT
Phenotype
MIM: 604806 MIM: 610400
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FLRT1 PubMed Links

Gene Details

Gene
FLRT1
Gene Name
fibronectin leucine rich transmembrane protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013280.4 Intron NP_037412.2
XM_005273861.1 Intron XP_005273918.1
XM_011544883.1 Intron XP_011543185.1
Gene
MACROD1
Gene Name
MACRO domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014067.3 Intron NP_054786.2
XM_005273939.4 Intron XP_005273996.1
XM_005273940.2 Intron XP_005273997.1
XM_006718521.3 Intron XP_006718584.1
XM_006718522.2 Intron XP_006718585.1
XM_011544970.2 Intron XP_011543272.1

View Full Product Details