Product Details

SNP ID
rs2336880
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.5:139684143 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATGCCTCAGAACCTGGACCTCCAGG[A/G]ACCTTCACTCACTTGGGGCCACCTC
Phenotype
MIM: 612752
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXXC5 PubMed Links

Gene Details

Gene
CXXC5
Gene Name
CXXC finger protein 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317199.1 164 Intron NP_001304128.1
NM_001317200.1 164 Intron NP_001304129.1
NM_001317201.1 164 Intron NP_001304130.1
NM_001317202.1 164 Intron NP_001304131.1
NM_001317203.1 164 Intron NP_001304132.1
NM_001317204.1 164 Intron NP_001304133.1
NM_001317205.1 164 Intron NP_001304134.1
NM_001317206.1 164 Intron NP_001304135.1
NM_001317207.1 164 Intron NP_001304136.1
NM_001317208.1 164 Intron NP_001304137.1
NM_001317209.1 164 Intron NP_001304138.1
NM_001317210.1 164 Intron NP_001304139.1
NM_001317211.1 164 Intron NP_001304140.1
NM_016463.8 164 Intron NP_057547.5
XM_017009573.1 164 Intron XP_016865062.1
Gene
PSD2
Gene Name
pleckstrin and Sec7 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032289.2 164 Intron NP_115665.1
XM_011537696.2 164 Intron XP_011535998.1
XM_017009975.1 164 UTR 5 XP_016865464.1
XM_017009976.1 164 Intron XP_016865465.1
XM_017009977.1 164 Intron XP_016865466.1

View Full Product Details