Product Details

SNP ID
rs34897167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:139687269 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCCCTGGACCTGACTCAGGACT[A/C]CAGACGAGGCCTTTGACCCTCTGCT
Phenotype
MIM: 612752
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CXXC5 PubMed Links

Gene Details

Gene
CXXC5
Gene Name
CXXC finger protein 5
There are no transcripts associated with this gene.

Gene
PSD2
Gene Name
pleckstrin and Sec7 domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032289.2 Intron NP_115665.1
XM_011537696.2 Intron XP_011535998.1
XM_017009975.1 Intron XP_016865464.1
XM_017009976.1 Intron XP_016865465.1
XM_017009977.1 Intron XP_016865466.1

View Full Product Details