Product Details
- SNP ID
-
rs11185518
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:196219664 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTGAGAGAGTGCAGCTGGTGGTA[C/T]AAGCCTTCCCTTACACAGCAGCTGT
- Phenotype
-
MIM: 612084
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC51A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs199580327] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC51A
- Gene Name
- solute carrier family 51 alpha subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_152672.5 |
|
Intron |
|
|
NP_689885.4 |
- Gene
- ZDHHC19
- Gene Name
- zinc finger DHHC-type containing 19
There are no transcripts associated with this gene.
View Full Product Details