Product Details

SNP ID
rs595241
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.12:133077993 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GTCTTTTCACCAGGCCAGTGAATTT[C/T]CTCTTTTTTTGCCCTCTTCCATTTT
Phenotype
MIM: 604082
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF140 PubMed Links

Gene Details

Gene
ZNF140
Gene Name
zinc finger protein 140
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300776.1 1449 Intron NP_001287705.1
NM_001300777.1 1449 Intron NP_001287706.1
NM_001300778.1 1449 Intron NP_001287707.1
NM_003440.3 1449 Intron NP_003431.2
XM_005266188.4 1449 Intron XP_005266245.1
XM_011534833.2 1449 Intron XP_011533135.1
XM_011534834.2 1449 UTR 5 XP_011533136.1
XM_011534835.2 1449 UTR 5 XP_011533137.1
XM_011534836.2 1449 Intron XP_011533138.1
XM_011534837.2 1449 Intron XP_011533139.1
XM_011534838.2 1449 Intron XP_011533140.1
XM_011534839.2 1449 Intron XP_011533141.1
XM_011534840.2 1449 Intron XP_011533142.1
XM_017019924.1 1449 Intron XP_016875413.1
XM_017019925.1 1449 UTR 5 XP_016875414.1

View Full Product Details