Product Details

SNP ID
rs924442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157751964 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCCTTAAGGACCAATTATTTTATCG[G/T]TTTAGTAATTTTTAGTTGATTTTCT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LSM11 PubMed Links

Gene Details

Gene
LSM11
Gene Name
LSM11, U7 small nuclear RNA associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173491.3 Intron NP_775762.1

View Full Product Details