Product Details

SNP ID
rs1875233
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.16:56652792 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CAGGCTTTCTGCTCTGAGCTCAGAT[A/G]GGTCAGGGCAGACTTTTTCTATTGG
Phenotype
MIM: 156349 MIM: 156352
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MT1B PubMed Links

Gene Details

Gene
MT1B
Gene Name
metallothionein 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005947.2 Intron NP_005938.1
Gene
MT1DP
Gene Name
metallothionein 1D, pseudogene
There are no transcripts associated with this gene.

Gene
MT1F
Gene Name
metallothionein 1F
There are no transcripts associated with this gene.

View Full Product Details